Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1H,1H,2H,2H-PERFLUOROOCTANE SULFONIC ACID 100 µG/ML IN METHANOL 1 * 1 mL
Catalog # EHERA15987125ME100
Supplier:
EHRENSTORFER
Expand image
1H,1H,2H,2H-PERFLUOROOCTANE SULFONIC ACID 100 µG/ML IN METHANOL 1 * 1 mL
Catalog # EHERA15987125ME100
Supplier:
EHRENSTORFER
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Calcium standard solution, 1000 mg/L Ca in 0,5 M nitric acid (from Ca(NO₃)₂) Certipur® standard for AAS, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW