ContactUsLinkComponent
Door with full sight glass window suitable for temperatures up to 300C 1 * 1 items
Catalog # MEMMB40574
Supplier:  MEMMERT
Door with full sight glass window suitable for temperatures up to 300C 1 * 1 items
Catalog # MEMMB40574
Supplier:  MEMMERT
Frequently Bought Together