1 component: Ammonium perfluoro(2-methyl-3-oxahexanoate) [CAS:62037-80-3] 100mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a freezer at -18C or below 1 * 5 Ampoul
Catalog # CMPLRD0998811.5
Supplier: CUSTOM MADE CHEMICALS LAB
1 component: Ammonium perfluoro(2-methyl-3-oxahexanoate) [CAS:62037-80-3] 100mg/l in Methanol CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store in a freezer at -18C or below 1 * 5 Ampoul
Catalog # CMPLRD0998811.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffer solutions, certified (DKD)
Supplier: Mettler - Toledo
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
VWR®, TOC standard solutions
Supplier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
Sodium benzoate, Supelco®
Supplier: Merck
pH buffer solutions, colour coded
Supplier: Hach