SWIVEL CASTOR LR POLY/ST RZ1/125 1 * 1 items
Catalog # HUPF0191445145
Supplier: HUPFER METALLWERKE
Frequently Bought Together
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: Hach
ABLUFTNADEL 832
Supplier: METROHM
GEAR KIT 1 * 1 KIT
Supplier: Cole Parmer
Accessories
Supplier: Brand
SPRAY ARM ADAPTER 1 * 1 items
Supplier: MIELE
hose set for gas connection complete 1 * 1 SET
Supplier: Binder
SERVICE KIT 2ND LEVEL P20L 1 * 1 items
Supplier: GILSON
Accessories for multi channel pipettes, mechanical, Research® plus
Supplier: EPPENDORF
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: Hach
POLYURETHANE TUBE DI 3.9 DO 6.1 BLUE 1 * 1 m
Supplier: EPPENDORF
SHAFT KIT 100, INCL.O-RING, FOR UHP SINGLE CH., L.BLUE 1 * 1 items
Supplier: VWR Collection
FD Control Assembly (includes PCB) 1 * 1 items
Supplier: LABCONCO
MOTOR FAN 1 * 1 items
Supplier: MEMMERT