ContactUsLinkComponent
Autoclave cleaner 10 sachets Special formulation for steam sterilizers. Cleans surface of stainless steel, copper, silicon rubber, plastic and similar dirt PLEASE NOTE: FOR EXTERNAL USE ONLY - NOT EDIBLE 1 * 1 SET
Catalog # HMCA54004
Supplier:  HMC Europe GmbH
Autoclave cleaner 10 sachets Special formulation for steam sterilizers. Cleans surface of stainless steel, copper, silicon rubber, plastic and similar dirt PLEASE NOTE: FOR EXTERNAL USE ONLY - NOT EDIBLE 1 * 1 SET
Catalog # HMCA54004
Supplier:  HMC Europe GmbH
Frequently Bought Together