ContactUsLinkComponent
RFM990-aus-32 digital refractometer with peltier temperature control. specifically configured and calibrated for measuring aqueous urea solutions in accordance with iso 22241. 1 * 1 items
Catalog # BELL19-73
Supplier:  BELLINGHAM STANLEY
RFM990-aus-32 digital refractometer with peltier temperature control. specifically configured and calibrated for measuring aqueous urea solutions in accordance with iso 22241. 1 * 1 items
Catalog # BELL19-73
Supplier:  BELLINGHAM STANLEY