ContactUsLinkComponent
The SLP-HS Single Reagent Set II is able to detect both peptidoglycan (PG) and (1->3)-β-D-glucan (BDG). The product uses a self-defe nse mechanism of silkworm Bombyx mori which leads to a color change by building a black melanin pigment in the presen 1 * 25 items
Catalog # WAKF296-81001
The SLP-HS Single Reagent Set II is able to detect both peptidoglycan (PG) and (1->3)-β-D-glucan (BDG). The product uses a self-defe nse mechanism of silkworm Bombyx mori which leads to a color change by building a black melanin pigment in the presen 1 * 25 items
Catalog # WAKF296-81001
Frequently Bought Together