ContactUsLinkComponent
Furnace MuFFle with scale anD soFtware l9/11/sw/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in t he rear wall 1 * 1 items
Catalog # NABELW095K1ANOTZ
Supplier:  NABERTHERM
Furnace MuFFle with scale anD soFtware l9/11/sw/c550 tMax1100 with FlaP Door anD over-teMPerature liMiter anD therMocouPle hole in t he rear wall 1 * 1 items
Catalog # NABELW095K1ANOTZ
Supplier:  NABERTHERM
Frequently Bought Together