ContactUsLinkComponent
workstation 24 in. x 24 in. x 33.8 in. with a 2.3 in. thick IG Series breadboard and I-125 Series pneumatic isolators that provide a 500 lb. gross load capacity. Mounting Holes 1/4-20 on 1.0in grid - *air compressor is required. 1 * 1 items
Catalog # ECHLNCVI2424
Supplier:  DISCOVER ECHO
workstation 24 in. x 24 in. x 33.8 in. with a 2.3 in. thick IG Series breadboard and I-125 Series pneumatic isolators that provide a 500 lb. gross load capacity. Mounting Holes 1/4-20 on 1.0in grid - *air compressor is required. 1 * 1 items
Catalog # ECHLNCVI2424
Supplier:  DISCOVER ECHO
Frequently Bought Together