Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66S3CE2I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66S3CE2I
Supplier: Sartorius Balances
Frequently Bought Together
Overhead stirrers, electronic, OHS digital and advance
Supplier: VELP SCIENTIFIC
Baths for immersion thermostats
Supplier: VWR Collection
Cole-Parmer® SHP-200 Series Large-Capacity Digital Magnetic Hotplate Stirrers
Supplier: Cole Parmer
Nalgene 30ml beaker rack for Solaris Shaker 11 x14 or 12 x14 Platforms, 36 places per rack 1 * 1 items
Supplier: Thermo Fisher Scientific
Dri-Block Aluminum Insert, 20 x 12 mm Vial, Shallow, Flat Bottom 1 * 1 items
Supplier: Cole-Parmer
Tubings, insulated
Supplier: LAUDA
Grinding balls for MM 200 and MM 400
Supplier: Retsch GmbH
Incubators, IF/IFplus (forced convection)
Supplier: MEMMERT
Precision PT100 reference sensors
Supplier: JULABO GmbH
Heating and drying ovens, Universal UN/UNplus (natural convection)
Supplier: MEMMERT
Dispersing elements for homogenisers
Supplier: IKA