Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66P3CF1I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 12 | 61 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA66P3CF1I
Supplier: Sartorius Balances
Frequently Bought Together
VWR® Standard 3500 Orbital Shaker
Supplier: VWR Collection
VWR® INCU-Line® Prime, Incubators with Natural Convection
Supplier: VWR Collection
VWR® VWB2, Water Baths
Supplier: VWR Collection
VWR® VV3, Vortex Mixer
Supplier: VWR Collection
Drying ovens with natural convection ED series, Avantgarde.Line
Supplier: Binder
Precision™ General purpose water baths
Supplier: THERMO ELECTRON LED
Orbital shakers, Solaris™ 2000 and 4000
Supplier: Thermo Fisher Scientific
Interchangeable blocks for block heaters, QBA, QBD and QBH
Supplier: GRANT INSTRUMENTS
Baths for immersion thermostats
Supplier: VWR Collection
Accessories for digesters DK series
Supplier: VELP SCIENTIFIC
Accessories for WTB Water Baths
Supplier: MEMMERT
Baths
Supplier: IKA
Ultrasonic liquid processors, programmable, Q500 and Q700
Supplier: QSONICA LLC
Accessories for Loopster basic and digital
Supplier: IKA
Stirrer bearings, PTFE
Supplier: Bohlender
Cole-Parmer® SH-200 See-Saw Rocking Shakers
Supplier: Cole Parmer
Bottle/tube rollers
Supplier: Thermo Fisher Scientific