Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S004I
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S004I
Supplier: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
VWR®, Digital Mini Block Heaters
Supplier: VWR Collection
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
VWR® VWB2, Water Baths
Supplier: VWR Collection
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
Findenser™ Air condenser for Magnetic Stirrer
Supplier: Heidolph Instruments GmbH & Co.KG
Gas lighter, piezoelectric
Supplier: USBECK Laborgeräte
VWR® VENTI-Line® Prime, Drying Ovens with Forced Convection
Supplier: VWR Collection
Magnetic stirrers, MST and MST Digital
Supplier: VELP SCIENTIFIC
VWR®, Mini vortex mixer
Supplier: VWR Collection
Hotplates, C-MAG HP series
Supplier: IKA
Burners
Supplier: USBECK Laborgeräte
Magnetic stirrer, Topolino
Supplier: IKA
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
Magnetic stirrer, Mini MR standard IKAMAG®
Supplier: IKA
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
Magnetic stirring bars
Supplier: VELP SCIENTIFIC
Accessories for Retsch sieves
Supplier: Retsch GmbH