ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S0034I
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer 1 * 1 items
Catalog # SARBMCA36S3S0034I
Supplier:  Sartorius Balances
Frequently Bought Together