ContactUsLinkComponent
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA36S3CF124H
Supplier:  Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
Catalog # SARBMCA36S3CF124H
Supplier:  Sartorius Balances
Frequently Bought Together