Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 mg, max. load 32 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA36S3CF
Supplier: Sartorius Balances
Frequently Bought Together
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Magnetic hotplate stirrer, RCT basic
Supplier: IKA
VWR®, Mini vortex mixer
Supplier: VWR Collection
Accessories for tube mill control
Supplier: IKA
VWR® VENTI-Line® Prime, Drying Ovens with Forced Convection
Supplier: VWR Collection
Replacement and spare parts for stirrer bearings, BOLA
Supplier: Bohlender
VWR® INCU-Line® IL 10, IL 23, IL 56, Digital Incubators
Supplier: VWR Collection
VWR® VMS-A, Magnetic Hotplate Stirrer
Supplier: VWR Collection
VWR® DRY-Line® Prime, Drying Ovens with Natural Convection
Supplier: VWR Collection
Vortex mixers, Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Gasprofi 1 SCS Micro Gas Burners
Supplier: WARTEWIG
VWR® lab dancer, Vortex Mixer
Supplier: VWR Collection
Cartridge 220G for cartridge burner
Supplier: USBECK Laborgeräte
VWR® VMS Advanced Magnetic Hotplate Stirrers
Supplier: VWR Collection
VWR® Standard 1000 Orbital Shaker
Supplier: VWR Collection