Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
SARBMCA36P3CF34H
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.001 | 0.01 mg, max. load 10.1 | 32 g. Draft Shield D & Activated Ionizer and Draft Shield Motors 1 * 1 items
SARBMCA36P3CF34H
Frequently Bought Together
VWR®, Accessories for DRY-Line, VENTI-Line and INCU-Line Prime
: VWR Collection
Disruptor Beads™ for Disruptor Genie®
: SCIENTIFIC INDUSTRIES ELMIS
VWR® Rocking Platform Shaker
: VWR Collection
VWR® lab dancer, Vortex Mixer
: VWR Collection
Stirring paddles, polypropylene
: KARTELL
Bunsen burner, cartridge type
: USBECK Laborgeräte
Immersion thermostats, CORIO™ C, CD and CP
: JULABO GmbH
Accessories for vortex mixers Vortex-Genie®
: SCIENTIFIC INDUSTRIES ELMIS
Tissue grinders, conical
: DWK Life Sciences
Shaking Water Baths, SW22 and SW23
: JULABO GmbH
Galvanised iron, wire gauzes with ceramic center for gas burners
: GLASWARENFABRIK KARL HECHT
Accessories for CR®, Viton® and reinforced tubings
: JULABO GmbH