Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA116S3S0013M
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D & Activated Draft Shield Motors 1 * 1 items
Catalog # SARBMCA116S3S0013M
Supplier: Sartorius Balances
Frequently Bought Together
Climate chambers, ICH series
Supplier: MEMMERT
Ø 9,52 mm x Ø 12,7 mm t=1,59mm aus virginalem PTF
Supplier: Bohlender
Heating mantle 2000ml 450W controlled temperature to 450C 1 * 1 items
Supplier: Cole Parmer
Accessories for overhead stirrer
Supplier: IKA
Accessories for Unheated Water Baths, Cole-Parmer®, WBU-200 Series
Supplier: Cole-Parmer
Bath, stainless steel, mixBATH
Supplier: 2MAG
Laboratory grinding mill, MultiDrive control
Supplier: IKA
Boiling baths, SBB Aqua Plus series
Supplier: GRANT INSTRUMENTS
Cryo-Compact circulators, CF series
Supplier: JULABO GmbH
Magnetic Stirrer, Fully Electronic
Supplier: GLASWARENFABRIK KARL HECHT
Perforated stainless steel shelf for sn 1 * 1 items
Supplier: MEMMERT