Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA116S3CF
Supplier: Sartorius Balances
Cubis® II High-Capacity Micro Balance. Scale div. (d) 0.002 mg, max. load 111 g. Draft Shield D 1 * 1 items
Catalog # SARBMCA116S3CF
Supplier: Sartorius Balances
Frequently Bought Together
Interchangeable blocks for PCMT and PCMT Plus Thermomixers
Supplier: GRANT INSTRUMENTS
Magnetic stirrers, C-MAG MS series
Supplier: IKA
Multi-position magnetic stirrers, MULTISTIRRER series
Supplier: VELP SCIENTIFIC
Stirring shafts, moon shaped, BOLA
Supplier: Bohlender
Accessories for knife mill, MICROTRON® MB 550
Supplier: KINEMATICA
Accessories for vortex mixers Vortex-Genie®
Supplier: SCIENTIFIC INDUSTRIES ELMIS
Accessories for ARE/AREX-6 digital and digital PRO hotplate stirrers
Supplier: VELP SCIENTIFIC
Orbital multi-platform shaker, PSU-20i
Supplier: GRANT INSTRUMENTS
Accessories for orbital shakers, KS 130
Supplier: IKA
Overhead stirrer tools, radial stirrers, six paddles, BOLA
Supplier: Bohlender
Grinding balls for planetary ball mills, PM 100, PM 100 CM, PM 200 and PM 400
Supplier: Retsch GmbH
Accessories for mixer mills MM 200 and MM 400
Supplier: Retsch GmbH
Blocks for IKA digital dry block heaters
Supplier: IKA
Dispersing vessels for Polytron dispersers
Supplier: KINEMATICA
Magnetic stirring bars, Komet
Supplier: THERMO H + P LABORTECHNIK
Orbital shaker, benchtop, MaxQ™ HP
Supplier: Thermo Fisher Scientific