DRUCKMINDERER FUER MEDICA PRO 1 * 1 ST
ELGAVAPSBR201264
Frequently Bought Together
p-Xylene 99% for spectroscopy
: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99.0-101.5% ACS
: Thermo Fisher Scientific
Acetone reagent ≥99.5% ACS
: Merck
Isoamyl alcohol 98%, pure
: Thermo Fisher Scientific
Ethylenediamine 99+%
: Thermo Fisher Scientific
Methanol, extra dry 99.9%, AcroSeal®
: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
: Thermo Fisher Scientific
Acetone 99.8% for HPLC
: Thermo Fisher Scientific
Cyclohexane, extra dry 99.5%, AcroSeal®
: Thermo Fisher Scientific
Water for analysis
: Thermo Fisher Scientific
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
: VWR Chemicals
Methanol ≥99.8% (by GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
: Honeywell Chemicals