ContactUsLinkComponent
Boric acid 40 g/l in water / tr NH4OH CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Catalog # CMPLZ12574494.1L
Boric acid 40 g/l in water / tr NH4OH CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15C to 25C 1 * 1 L
Catalog # CMPLZ12574494.1L
Frequently Bought Together