Capacitive rod sensor, housing material: PTFE, thread: M12x1, length sensor: 70mm with LED display, length cable: 2 m, for ATEX zon e 0, ATEX Ex II 1G Ex ia IIC T1-T6 Ga, output function: NAMUR, operating distance 1 - 6 mm adjustable, Switch amplifi 1 * 1 items
Catalog # SCTV108277
Supplier: S.C.A.T.
Frequently Bought Together