Exhaust filter L, V3.0, economy package, with splash guard and change label, service life 12 months, quantity = 2 pieces 1 * 2 items
Catalog # SCTV490915
Supplier: S.C.A.T.
Exhaust filter L, V3.0, economy package, with splash guard and change label, service life 12 months, quantity = 2 pieces 1 * 2 items
Catalog # SCTV490915
Supplier: S.C.A.T.
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Accessories for SCAT Safety Caps
Supplier: S.C.A.T.
SafetyWasteCaps
Supplier: S.C.A.T.
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Jerricans, narrow neck, HDPE, UN Approved
Supplier: BURKLE
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals