ContactUsLinkComponent
Exhaust filter M, V3.0, economy package, with splash guard and change indicator, box of 20 pieces 1 * 20 items
Catalog # SCTV490337
Supplier:  S.C.A.T.
undefined
Exhaust filter M, V3.0, economy package, with splash guard and change indicator, box of 20 pieces 1 * 20 items
Catalog # SCTV490337
Supplier:  S.C.A.T.
Frequently Bought Together