ContactUsLinkComponent
8 components: 1000mg/l ICP Standard ,, 8 components: 1000mg/l each of Cr Cu Co Cd Ni Pb Zn Mo in HNO3 2% CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months. (12 months after opening),,,, ,,Store under normal laboratory conditions 1 * 100 mL
Catalog # CMPLRM098902L1
8 components: 1000mg/l ICP Standard ,, 8 components: 1000mg/l each of Cr Cu Co Cd Ni Pb Zn Mo in HNO3 2% CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months. (12 months after opening),,,, ,,Store under normal laboratory conditions 1 * 100 mL
Catalog # CMPLRM098902L1
Frequently Bought Together