ContactUsLinkComponent
Micro electrode, temperature sensor Pt 1000, 3 m cable with BNC and banana plug, glass shaft, platinum diaphragm, electrolyte KCl 3 mol/l, Silamid lead with silver ion barrier and double electrolyte, cylinder membrane, A-glass, length 70/130 mm, 12/ 1 * 1 items
Catalog # SCOI285101340
Supplier:  SI Analytics
Micro electrode, temperature sensor Pt 1000, 3 m cable with BNC and banana plug, glass shaft, platinum diaphragm, electrolyte KCl 3 mol/l, Silamid lead with silver ion barrier and double electrolyte, cylinder membrane, A-glass, length 70/130 mm, 12/ 1 * 1 items
Catalog # SCOI285101340
Supplier:  SI Analytics
Frequently Bought Together