Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
(3RS)-9-Fluoro-3-methyl-10-(4-methyl-4-oxidopiperazin-1-yl)-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid certified reference material, TraceCERT(R) 1 * 100 mg
PHR9018-100MG
:
Merck
(3RS)-9-Fluoro-3-methyl-10-(4-methyl-4-oxidopiperazin-1-yl)-7-oxo-2,3-dihydro-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid certified reference material, TraceCERT(R) 1 * 100 mg
PHR9018-100MG
:
Merck
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
Butane-1,3-diol, Reference Standard
: USP
Isomaltulose, Reference Standard
: USP
CEFTRIAXONE IMPURITY A 1 * 10 mg
: EDQM
Bis(2-ethylhexyl)phthalate, Supelco®
: Merck
100mg/l each of As,Be,Bi,Ca,Cd,Co,Cr,Cu,Fe,Li,Mg,Mn,Mo,Ni,Pb,Sb,Se,Sr,Ti,Tl,V,Zn 1 * 500 mL
: CUSTOM MADE CHEMICALS LAB
Buffer solutions in sachets, perpHect® Pouches, Orion™
: Thermo Orion
Nafcillin, Reference Standard
: USP
Rotigotine Related Compounds, Reference Standard
: USP
Clean Clay Loam, (CRM), Supelco®
: Merck
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
: Honeywell Chemicals
ARMEPAVINE, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
: Merck
16Alpha-Hydroxy Prednisolone-9(11)-ene, Pharmaceutical Analytical Impurities
: USP
Pantoprazole Related Compound C pharmaceutical secondary standard, certified reference material 1 * 50 mg
: Merck
Supelco® Organic Reference Standard, Chrysene
: Merck
Supelco® Organic Reference Standard, Isopentyl acetate
: Merck
Organic reference standard, 4-Methoxyphenol, analytical standard, Supelco®
: Merck
PHOSPHATE BUFFERED SALINE, PH 7.4, PHAST 1 * 20 items
: Merck