Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,5-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazolium nitrate certified reference material, Trace CERT(R) 1 * 100 mg
Catalog # PHR9001-100MG
Supplier:
Merck
Expand image
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[[(2,5-dichlorophenyl)methyl]oxy]ethyl]-1H-imidazolium nitrate certified reference material, Trace CERT(R) 1 * 100 mg
Catalog # PHR9001-100MG
Supplier:
Merck
Frequently Bought Together
Supelco® PESTANAL® Organic Reference Standard, Chlorothalonil
Supplier: Merck
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
Pimecrolimus
Supplier: USP
AVENANTHRAMIDE D, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Supplier: Merck
Sulfamethoxazole-(phenyl-13C6), analytical standard, VETRANAL™, Supelco®
Supplier: Merck
Dienochlor certified 1 * 0,25 g
Supplier: EHRENSTORFER
Calcium Magnesium Citrate, Reference Standard
Supplier: USP
Supelco® Organic Reference Standard, Lycopene
Supplier: Merck
FENPROPIMORPH 1 * 250 mg
Supplier: EHRENSTORFER
Single element standards for ICP-OES, copper
Supplier: Agilent
Supelco® Organic Reference Standard, (+)-Menthofuran
Supplier: Merck
Cevimeline Related Compounds, Reference Standard
Supplier: USP
Dibutyl Sebacate, Reference Standard
Supplier: USP
PERFLUORO-3,6-DIOXAHEPTANOIC ACID 50 µG/ML IN METHANOL:WATER 1 * 1 mL
Supplier: EHRENSTORFER
5,7-DIHYDROXY-4-METHYLCOUMARIN, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 100 mg
Supplier: Merck
TOLUENE D8 1 * 0,1 g
Supplier: EHRENSTORFER
Pharmaceutical secondary standard, trimeprazine tartrate, (CRM), Supelco®
Supplier: Merck