Bendamustine Hydrochloride Monohydrate certified reference material, pharmaceutical secondary standard 1 * 500 mg
Catalog # PHR2333-500MG
Supplier: Merck
Bendamustine Hydrochloride Monohydrate certified reference material, pharmaceutical secondary standard 1 * 500 mg
Catalog # PHR2333-500MG
Supplier: Merck
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Silicone Rotational viscosity standards
Supplier: VWR Chemicals
Buffer solutions
Supplier: Hanna
VWR® ARISTAR®, Antimony Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Colour calibration standard solutions, Pt-Co, Supelco®
Supplier: Merck
Amitrole 10 ng/µl 1 * 10 mL
Supplier: EHRENSTORFER
VWR®, Succinate 1000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
n-Decane, Supelco®
Supplier: Merck
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals