Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-Propanol Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1208-3X1.2M
Supplier:
Merck
Expand image
1-Propanol Pharmaceutical Secondary Standard Certified Reference Material 1 * 3 items
Catalog # PHR1208-3X1.2M
Supplier:
Merck
Frequently Bought Together
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Hydrochloric acid 1,128% (w/v) analytical reagent
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Density Standards for the Petroleum Industry, Certified Reference Materials
Supplier: VWR Chemicals