CHECKIT® Testpak Chlorine DPD, test range 0 - 1 mg/L Cl2. Supplied field-ready in a carrying case with reagents and accessories for 30 tests 1 * 1 KIT
Catalog # LOVI147510
Supplier: Lovibond
CHECKIT® Testpak Chlorine DPD, test range 0 - 1 mg/L Cl2. Supplied field-ready in a carrying case with reagents and accessories for 30 tests 1 * 1 KIT
Catalog # LOVI147510
Supplier: Lovibond
Frequently Bought Together
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
VWR® Cryolab Q Path, Freezing Aerosol
Supplier: VWR Chemicals
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach