ContactUsLinkComponent
The Allergen Rapid Test EASY is intended for the rapid detection of allergen ingredients on surrounding surfaces (swab sample) and in rinse water samples. It is a qualitative test in lateral flow immunoassay format for visual detection. 1 * 10 Tests
Catalog # SCIL1500017
Supplier:  HYSERVE
The Allergen Rapid Test EASY is intended for the rapid detection of allergen ingredients on surrounding surfaces (swab sample) and in rinse water samples. It is a qualitative test in lateral flow immunoassay format for visual detection. 1 * 10 Tests
Catalog # SCIL1500017
Supplier:  HYSERVE
Frequently Bought Together