Immunotag CD45 (LCA) (PT1659) mouse mAb, Alexa Fluor® 555 1 * 100 µG
Catalog # GENOITM6018100U555
Supplier: G-Biosciences
Immunotag CD45 (LCA) (PT1659) mouse mAb, Alexa Fluor® 555 1 * 100 µG
Catalog # GENOITM6018100U555
Supplier: G-Biosciences
Frequently Bought Together
Transfer pipettes
Supplier: RATIOLAB
Petri dishes
Supplier: Anicrin
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Multiple well cell culture plates, Costar®
Supplier: Corning
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF