Immunotag CD45 (LCA) (PT1659) mouse mAb, Alexa Fluor® 488 1 * 100 µG
Catalog # GENOITM6018100U488
Supplier: G-Biosciences
Immunotag CD45 (LCA) (PT1659) mouse mAb, Alexa Fluor® 488 1 * 100 µG
Catalog # GENOITM6018100U488
Supplier: G-Biosciences
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Sampling scoops, LaboPlast®
Supplier: BURKLE
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Individually Wrapped Serological Pipettes, Falcon®
Supplier: Corning
Desiccant pack, Whatman™
Supplier: Whatman products (Cytiva)
UV microplates, 96-well, Corning®
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
Detectable spoons, SteriPlast®
Supplier: BURKLE
Sampling scoops, long handle, disposable, LaboPlast®/SteriPlast®
Supplier: BURKLE
TILT bottles, DURAN®
Supplier: DWK Life Sciences