Pyrrolizidine alkaloids Mixture 661 100 µg/mL in Methanol 1 * 1 mL
Catalog # EHERA50000661M
Supplier: EHRENSTORFER
Pyrrolizidine alkaloids Mixture 661 100 µg/mL in Methanol 1 * 1 mL
Catalog # EHERA50000661M
Supplier: EHRENSTORFER
Frequently Bought Together
2,6-Dichlorobenzamide
Supplier: EHRENSTORFER
Gold standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
Organic reference standard, Diethylnitrosoamine 100 µg/ml in methanol, Dr Ehrenstorfer
Supplier: EHRENSTORFER
[EN]ROTARY VANE PUMP FLUID, AVF 60 GOLD 1 * 1 L
Supplier: Agilent
Supelco® VETRANAL® Organic Reference Standard, α-Estradiol
Supplier: Merck
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
Imibenconazole certified 1 * 0,1 g
Supplier: EHRENSTORFER
Amaranth certified 1 * 0,25 g
Supplier: EHRENSTORFER