ContactUsLinkComponent
Nitrates (NO3-) 1000mg/l Chlorides (Cl-) 1000mg/l Sulphates (SO42-) 1000mg/l Bromides (Br-) 100mg/l Fluorides (F-) 100mg/l Phosphates (PO43-) 100mg/l Nitrites (NO2-) 50mg/l in Water 1 * 100 mL
Catalog # CMPLRM056759L1
Nitrates (NO3-) 1000mg/l Chlorides (Cl-) 1000mg/l Sulphates (SO42-) 1000mg/l Bromides (Br-) 100mg/l Fluorides (F-) 100mg/l Phosphates (PO43-) 100mg/l Nitrites (NO2-) 50mg/l in Water 1 * 100 mL
Catalog # CMPLRM056759L1
Frequently Bought Together