ContactUsLinkComponent
TABLE MOB + SHELF & UPSTAND 600X600 Dimensions Width: 600mm Depth: 600mm Height: 840mm Floor clearance to undershelf: 300mm Specification Fully welded 304-grade stainless steel Brushed dull polish finish 1.5mm thick stainless steel sheet 4 x stainles 1 * 1 items
Catalog # TEKMTLTA1SUPM60X60
Supplier:  TEKNOMEK
TABLE MOB + SHELF & UPSTAND 600X600 Dimensions Width: 600mm Depth: 600mm Height: 840mm Floor clearance to undershelf: 300mm Specification Fully welded 304-grade stainless steel Brushed dull polish finish 1.5mm thick stainless steel sheet 4 x stainles 1 * 1 items
Catalog # TEKMTLTA1SUPM60X60
Supplier:  TEKNOMEK
Frequently Bought Together