ContactUsLinkComponent
12 components: 100mg/l each of Al Ba B Co Cr Fe P Mn Ni Pb Cu Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 1 * 100 mL
Catalog # CMPLRM097127L1
12 components: 100mg/l each of Al Ba B Co Cr Fe P Mn Ni Pb Cu Zn in HNO3 2% CRM, ISO 17034 and ISO 17025 Conservation 24 months. (12 months after opening) Store under normal laboratory conditions, at temperatures between 15C to 1 * 100 mL
Catalog # CMPLRM097127L1
Frequently Bought Together