ContactUsLinkComponent
Spline Kit, 10mm diameter x 150mm, aluminum Light weight spline shaft kit (10mm dia. x 150mm, aluminum, 43g shaft). Use to reduce base load of Taber Linear Abraser includes spline shaft and bearing. 1 * 1 KIT
Catalog # SIMETB0000-132280
Supplier:  Industrial Physics
Spline Kit, 10mm diameter x 150mm, aluminum Light weight spline shaft kit (10mm dia. x 150mm, aluminum, 43g shaft). Use to reduce base load of Taber Linear Abraser includes spline shaft and bearing. 1 * 1 KIT
Catalog # SIMETB0000-132280
Supplier:  Industrial Physics
Frequently Bought Together