Internal PCR control with RNA template and VIC probe 1 * 150 Tests
Catalog # PRDEZ-INT-RNA-VIC
Supplier: PRIMERDESIGN
Internal PCR control with RNA template and VIC probe 1 * 150 Tests
Catalog # PRDEZ-INT-RNA-VIC
Supplier: PRIMERDESIGN
Frequently Bought Together
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Supplier: OMEGA BIO-TEK
2-Propanol ≥99% for biotechnology
Supplier: VWR Chemicals
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
illustra™ TempliPhi™100/500 Amplification Kits
Supplier: Cytiva
Ultra-Plex™ 1-Step ToughMix® (4X)
Supplier: Quantabio
jetPEI® HTS DNA Transfection Reagents
Supplier: SARTORIUS
IPTG (isopropyl-β-D-thiogalactopyranoside) for molecular biology dioxane-free
Supplier: PanReac AppliChem
PROCHO5 W/PLURONIC W/O L-GLN OR HT 1 * 1 L
Supplier: LONZA
Lysing kits for Fastprep®-24 5G, Lysing Matrix E, MP Biomedicals
Supplier: MP Biomedicals