ContactUsLinkComponent
The Pierce High-Capacity Protein A Magbeads Alkali Stable Is A High-Performance Magnetic Bead For Purifying Antibodies From Cell Cul ture Supernatant Ascites Serum And Crude Extracts. The Protein A Ligand Has Been Genetically Modified To Provide Stab 1 * 25 mL
Catalog # PIERA53037
undefined
The Pierce High-Capacity Protein A Magbeads Alkali Stable Is A High-Performance Magnetic Bead For Purifying Antibodies From Cell Cul ture Supernatant Ascites Serum And Crude Extracts. The Protein A Ligand Has Been Genetically Modified To Provide Stab 1 * 25 mL
Catalog # PIERA53037
Frequently Bought Together