ContactUsLinkComponent
20 components: Al 2.5mg/l As 2.5mg/l Ba 2.5mg/l Be 0.5mg/l B 5mg/l Cd 0.5mg/l Cr 2mg/l Co 1mg/l Cu 2mg/l Pb 2.5mg/l Mn 2.5mg/l Ni 2mg/l P 25mg/l Ag 1mg/l Sb 2.5mg/l Sr 2.5mg/l U 2.5mg/l Zn 5mg/l Fe 30mg/l Rh 12.5 1 * 100 mL
Catalog # CMPLRM012187L1
20 components: Al 2.5mg/l As 2.5mg/l Ba 2.5mg/l Be 0.5mg/l B 5mg/l Cd 0.5mg/l Cr 2mg/l Co 1mg/l Cu 2mg/l Pb 2.5mg/l Mn 2.5mg/l Ni 2mg/l P 25mg/l Ag 1mg/l Sb 2.5mg/l Sr 2.5mg/l U 2.5mg/l Zn 5mg/l Fe 30mg/l Rh 12.5 1 * 100 mL
Catalog # CMPLRM012187L1