2-Dicyclohexylphosphino-2,4,6-tri-iso 1 * 100 g
Catalog # ABCRAB148085_100
Supplier: abcr
Frequently Bought Together
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Acetonitrile ≥99.8% for HPLC
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific
Toluene, extra dry 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Acetone ≥99.5% NF, FCC, J.T.Baker®
Supplier: Avantor
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute (untaxed) ≥99.8% (by GC), CHROMASOLV™ A15 for HPLC, without additive, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific