50 TABLETS MOLYBDATE LR N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHMDL150
Supplier: WATER ID PRIMELAB
50 TABLETS MOLYBDATE LR N° 1 PHOTOMETER 1 * 50 items
Catalog # WTIDTBSHMDL150
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Test kits, chlorine, Spectroquant®, Supelco®
Supplier: Merck
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach
pH indicators with integral comparison strips, Whatman™
Supplier: Whatman products (Cytiva)
Immersion oil
Supplier: ZEISS