REAGENT KIT FOR PRIMELAB ID66(250 TESTS) 1 * 1 items
Catalog # WTIDPL66-KIT
Dodavatel: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID66(250 TESTS) 1 * 1 items
Catalog # WTIDPL66-KIT
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
Test strips for peroxide, MQuant®, Supelco®
Dodavatel: Merck
Sodium hydroxide 97.0-100.5%, pellets NF, JP, Ph. Eur., BP
Dodavatel: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Dodavatel: Merck
Visual colour kits, ammonium, MQuant®, Supelco®
Dodavatel: Merck
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Dodavatel: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Formalin 10%, Q Path®, buffered
Dodavatel: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Dodavatel: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Dodavatel: Merck
Test strips for copper, MQuant®, Supelco®
Dodavatel: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Dodavatel: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Dodavatel: VWR Chemicals
Test kits, photometric, phosphate, LCK cuvette tests
Dodavatel: Hach