REAGENT KIT FOR PRIMELAB ID166(50 TESTS) 1 * 1 items
Catalog # WTIDPL166-KIT
Supplier: WATER ID PRIMELAB
REAGENT KIT FOR PRIMELAB ID166(50 TESTS) 1 * 1 items
Catalog # WTIDPL166-KIT
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Sodium hydroxide ≥98%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, nitrite, MQuant®, Supelco®
Supplier: Merck
Papanicolaou's solution 2a Orange G (OG 6), Sigma-Aldrich®
Supplier: Merck
Visual colour kits, iron, MQuant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test Strips, nitrate, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach