100 X 25 TUBETESTS COD LR (0 - 150 MG/L) 1 * 2.500 Vial
Catalog # WTIDCOD-79-LR-100
Supplier: WATER ID PRIMELAB
100 X 25 TUBETESTS COD LR (0 - 150 MG/L) 1 * 2.500 Vial
Catalog # WTIDCOD-79-LR-100
Supplier: WATER ID PRIMELAB
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Visual colour kits, chlorine, MQuant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test strips for nickel, MQuant®, Supelco®
Supplier: Merck
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
VWR®, OCT Embedding matrix for frozen sections
Supplier: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium hydroxide ≥97.0%, shot, TECHNICAL
Supplier: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Zinc, LCK and LCS cuvette tests
Supplier: Hach
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach
Nitrite VHR L, 500 ml
Supplier: Lovibond
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Nitrate, LCK cuvette tests
Supplier: Hach