200 X 25 TUBETESTS COD HR (0-15000 MG/L) 1 * 5.000 Vial
Catalog # WTIDCOD-17-HR-200
Supplier: WATER ID PRIMELAB
200 X 25 TUBETESTS COD HR (0-15000 MG/L) 1 * 5.000 Vial
Catalog # WTIDCOD-17-HR-200
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Visual colour kits, ammonium, MQuant®, Supelco®
Supplier: Merck
Test kits, TOC, Spectroquant®, Supelco®
Supplier: Merck
Sodium hydroxide solution 10%, AnalaR NORMAPUR®
Supplier: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Ammonia solution 25%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Test kits, photometric, AccuVac®
Supplier: Hach
Test kits, digital titration method
Supplier: Hach
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
Test cassettes for urine
Supplier: SURESCREEN
VISOCOLOR® Colorimetric Test, Cyanuric acid
Supplier: MACHEREY-NAGEL