E.E. MOSSEL DC TUBES 1 * 20 items
760304WA
Frequently Bought Together
VWR® Culture Media, Ready to Use, in Bags
: VWR Chemicals
VWR® Culture Media, Ready To Use, In Tubes
: VWR Chemicals
Dehydrated culture media
: Merck
VWR® Scholten's Dehydrated Culture Medium
: VWR Chemicals
Media supplements for dehydrated culture media
: THERMO FISHER DIAGNOSTICS
Viabank™ Bacterial Storage Beads
: MEDICAL WIRE EQUIPMENT
POTATO DEXTROSE AGAR 1 * 500 g
: LIOFILCHEM
VWR® Ready to Use Media Plates (90 mm), Irradiated
: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
: VWR Chemicals
FASSIF BUFFER CONCENTRATE (6L) 250G 1 * 250 g
: MARKET SOURCE PART PROCESS
VWR®, Pre-weighed dehydrated culture media in sachets
: VWR Chemicals