HOSE PUMP PLP381 BEHROTEST LABORATORY 1 * 1 items
Catalog # BEHRB00734903
Supplier: BEHR
Frequently Bought Together
SmartBlocks for ThermoMixer® C and ThermoStat C
Supplier: EPPENDORF
Single gas detectors, BW Clip
Supplier: BW TECHNOLOGIES
Gas detection tube for hydrogen sulphide
Supplier: Dräger
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Air flow indicator
Supplier: Dräger
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Plastic cuvette for LICO
Supplier: Hach
Central connector for absorption vessel ASH 1 * 1 items
Supplier: BEHR
Accessories for Reaction Vessels
Supplier: KGW
Accessories for ÄKTA chromatography systems
Supplier: Cytiva
Storage phosphor screens and cassettes
Supplier: Cytiva