Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BARIUM CHLORIDE DIHYDRATE ACS REAGENT 1 * 500 g
Catalog # SIAL217565-500G
Supplier:
Merck
Expand image
BARIUM CHLORIDE DIHYDRATE ACS REAGENT 1 * 500 g
Catalog # SIAL217565-500G
Supplier:
Merck
Frequently Bought Together
Nickel (II) oxide, Technipur®
Supplier: Merck
Potassium dichromate ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Mercury(II) chloride 99.5%
Supplier: Thermo Fisher Scientific
Gallium(III) chloride, anhydrous 99.999%
Supplier: Apollo Scientific
Sodium borohydride ≥98%
Supplier: Thermo Fisher Scientific
Sodium nitrate 99+% ACS
Supplier: Thermo Fisher Scientific
Iron(III) oxide 95%, pure
Supplier: Thermo Fisher Scientific
Aluminium chloride, anhydrous 99%, granules, extra pure
Supplier: Thermo Fisher Scientific